Learn More
World Precision Instrument Potassium Ion Exchanger
Make micropipettes to record cellular concentrations
WPI’s Liquid Ion Exchangers (LIX), for use with the FD223A Electrometer, allow intracellular measurements to be made for cations (hydrogen, potassium and calcium).