Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Nitrate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
:
1 g
Specifications
Specifications
CAS | 7757-79-1 |
Quantity | 1 g |
Type | Certified Reference Material |
Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
Linear Formula | KNO3 |
MDL Number | MFCD00011409 |
Synonym | Potassium nitrate |
Formula Weight | 101.10 |
Grade | Certified Reference Material |
Melting Point | 334°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction