Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Potassium Nitrite
SCI_ED

Catalog No. p-4416433 Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Reagent Grade
This item is not returnable. View return policy
This item is not returnable. View return policy

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7758-09-0
Molecular Formula KNO2
Molecular Weight (g/mol) 85.10
MDL Number MFCD00011408
InChI Key BXNHTSHTPBPRFX-UHFFFAOYSA-M
Synonym potassium nitrite, nitrous acid, potassium salt, caswell no. 698, ccris 3959, potassium nitrite 1:1, epa pesticide chemical code 076203, nitrous acid, potassium salt 1:1, kaliumnitrit, acmc-1bi01, dsstox_cid_22320
PubChem CID 516910
IUPAC Name potassium nitrite
SMILES [K+].[O-]N=O

Specifications

Color White to Yellow
Quantity 500 g
Assay Percent Range >96%
Linear Formula KNO2
Formula Weight 85.11
Grade Reagent
Packaging Reagent Grade
Density 1.915g/mL
Chemical Name or Material Potassium Nitrite
ShelfLife 2 years
DOTInformation DOT Class 5.1, II, : Oxidizer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.