Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Potassium Permanganate
SCI_ED

Catalog No. p-4416434 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Reagent Grade

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7722-64-7
Molecular Formula KMnO4
Molecular Weight (g/mol) 158.032
InChI Key VZJVWSHVAAUDKD-UHFFFAOYSA-N
Synonym potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
PubChem CID 516875
IUPAC Name potassium;permanganate
SMILES [O-][Mn](=O)(=O)=O.[K+]

Specifications

Color Brown
Quantity 100 g
Assay Percent Range >95%
Linear Formula KMnO4
Formula Weight 158.04
Packaging Reagent Grade
Chemical Name or Material Potassium Permanganate
ShelfLife 5 years
DOTInformation DOT Class 5.1, II, : Oxidizer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.