Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Permanganate, 89.1mg/L Equivalent to 100ppm Chlorine, Reagents

Catalog No. 01334125
Click to view available options
:
Quantity: 500 mL

Potassium Permanganate, 89.1mg/L Equivalent to 100ppm Chlorine, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5, 7722-64-7
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Packaging Glass Bottle
Quantity 500 mL
Color Purple
For Use With (Equipment) Water Analyzer
Percent Purity 0.009%
Grade Reagent
Boiling Point 100°C
Specific Gravity 1.0001
Solubility Completely miscible
Physical Form Liquid
Chemical Name or Material Potassium Permanganate, 891 mg/L Equivalent to 1000 ppm Chlorine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.