Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Permanganate, For Mercury, Metals, Certified, 5.0% (w/v) ±0.5%, LabChem™

Catalog No. LC199404
Click to view available options
Quantity:

Chemical Identifiers

CAS

7722-64-7, 7732-18-5

Molecular Formula

KMnO4

Molecular Weight (g/mol)

158.032

InChI Key

VZJVWSHVAAUDKD-UHFFFAOYSA-N

Synonym

potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo

PubChem CID

516875

IUPAC Name

potassium;permanganate

SMILES

[O-][Mn](=O)(=O)=O.[K+]

Specifications

CAS Max %

95, 5

Linear Formula

KMnO4

Density

1g/mL

Formula Weight

158.04

Physical Form

Liquid

Percent Purity

5.0% ± 0.5% (w/v)

Grade

Certified

Identification

Passes Test

Packaging

Amber Glass

Solubility

Soluble in water

Specific Gravity

1g/mL

Storage Information

Room Temperature

Traceability to NIST

Traceable to NIST

Decomposition Information

Manganese; Oxygen

Color

Purple

Quantity

4L

Chemical Name or Material

Potassium Permanganate, For Mercury, Metals

Hazard Statement

GHS H Statement
Toxic to aquatic life with long lasting effects.

Precautionary Statement

GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.