Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium permanganate solution, Volumetric, 0.1 M KMnO4 (0.5N), Honeywell Fluka™

Catalog No. 60046542 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
Packaging:

Volumetric, 0.1 M KMnO4 (0.5N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

Chemical Identifiers

CAS

7722-64-7, 7732-18-5

Molecular Formula

KMnO4

Molecular Weight (g/mol)

158.032

MDL Number

MFCD00011364

InChI Key

VZJVWSHVAAUDKD-UHFFFAOYSA-N

Synonym

potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo

PubChem CID

516875

IUPAC Name

potassium;permanganate

SMILES

[O-][Mn](=O)(=O)=O.[K+]

Specifications

Boiling Point

100°C

CAS Min %

≥90.0000%, ≥0.2500%

CAS Max %

≤100.0000%, <2.5000%

Quantity

4 x 2.5 L

Linear Formula

KMnO4

UN Number

UN3082

Formula Weight

158.03

Concentration or Composition (by Analyte or Components)

0.1 M KMnO4 (0.5N)

Packaging

Glass Bottle

Chemical Name or Material

Potassium permanganate solution, Volumetric, 0.1 M KMnO4 (0.5N)

Hazard Statement

P273

ShelfLife

540 days from date of manufacture

DOTInformation

9 / PGIII

Recommended Storage

Room Temp

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.