Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Potassium Phosphate Monobasic
SCI_ED

Catalog No. p-4416437 Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
Packaging:

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS

7778-77-0

Molecular Formula

H2KO4P

Molecular Weight (g/mol)

136.08

MDL Number

MFCD00011401 MFCD00147253

InChI Key

GNSKLFRGEWLPPA-UHFFFAOYSA-M

Synonym

potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate

PubChem CID

516951

ChEBI

CHEBI:63036

IUPAC Name

potassium dihydrogen phosphate

SMILES

[K+].OP(O)([O-])=O

Specifications

Color

White

Quantity

100 g

Assay Percent Range

99% w/v

Linear Formula

KH2PO4

Formula Weight

136.09

Packaging

Solid

Chemical Name or Material

Potassium Phosphate, Monobasic

ShelfLife

2 years

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.