Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education™ Potassium Phosphate, Tribasic
Shop All AquaPhoenix Scientific Products

Click to view available options
Quantity:
100 g
500 g
Packaging:
Reagent Grade
Description
This product is for educational use - no C of A is available.
Specifications
Specifications
Color | White |
Quantity | 100 g |
Assay Percent Range | >95% |
Linear Formula | K3PO4·nH2O |
Formula Weight | 212.03 |
Packaging | Reagent Grade |
Chemical Name or Material | Potassium Phosphate, Tribasic |
Safety and Handling

ShelfLife | 2 years |
DOTInformation | DOT Class 8, II, : Corrosive |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction