Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Rhodizonate 95.0+%, TCI America™
SDP

Catalog No. R00581G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 13021-40-4
Molecular Formula C6H2KO6
Molecular Weight (g/mol) 209.17
MDL Number MFCD00001649
InChI Key UGFHFZSZCJFOTO-UHFFFAOYSA-N
Synonym potassium rhodizonate, rhodizonic acid dipotassium salt, dipotassium rhodizonate, 5-cyclohexene-1,2,3,4-tetrone, 5,6-dihydroxy-, dipotassium salt, potassium 3,4,5,6-tetraoxocyclohex-1-ene-1,2-bis olate, 5-cyclohexene-1,2,3,4-tetrone, 5,6-dihydroxy-, potassium salt 1:2, dipotassium 3,4,5,6-tetraoxocyclohexene-1,2-diolate, kaliumrhodizonat, dipotassium tetraoxocyclohex-1-ene-1,2-bis olate, potassium rhodizonate, technical grade
PubChem CID 83051
IUPAC Name dihydroxycyclohex-5-ene-1,2,3,4-tetrone potassium
SMILES [K].OC1=C(O)C(=O)C(=O)C(=O)C1=O

Specifications

Color Black
Quantity 1 g
Formula Weight 246.26
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium Rhodizonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.