Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Sodium Tartrate, Tetrahydrate, Purified, 95%, Spectrum™ Chemical
SDP

Catalog No. 18605710 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
Packaging:

P1401, 6381-59-5, C4H4KNaO6•4H2O

Spectrum™ Chemical Potassium Sodium Tartrate, Tetrahydrate, Purified was one of the first materials discovered to exhibit piezoelectricity. It is a double salt and is deliquescent so the material deteriorates if stored in damp conditions The Purified Grade describes chemicals of good quality where there are no official standards and is normally limited to inorganic chemicals.

Chemical Identifiers

CAS

6381-59-5

Molecular Formula

C4H12KNaO10

Molecular Weight (g/mol)

282.22

InChI Key

VZOPRCCTKLAGPN-ZFJVMAEJSA-L

IUPAC Name

potassium sodium (2R,3R)-2,3-dihydroxybutanedioate tetrahydrate

SMILES

O.O.O.O.[Na+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

Specifications

CAS Max %

1

Packaging

Fiber Drum

Quantity

50 kg

Percent Purity

95%

Grade

Purified

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.