Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Sorbate, MilliporeSigma™ Supelco™

Catalog No. 111013684 Shop All MilliporeSigma Supelco Products
Click to view available options
:
1,000 mg

Potassium sorbate is a preservative, commonly used in a variety of food and beverages. It is a food antimicrobial agent, which can act effectively along a wide pH range to inhibit the growth of yeast, molds and some bacteria.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Packaging Ampule
Quantity 1000 mg
CAS 24634-61-5
Type Food Additive Standard
Linear Formula C6H7 KO2
MDL Number MFCD00016546
Synonym Sorbic acid potassium salt; 2,4-Hexadienoic acid potassium salt
Formula Weight 150.22
Grade Analytical Standard
Physical Form Neat
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.