Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Standard, 100 ppm APHA, Reagents

Catalog No. 01334241
Click to view available options
:
Quantity: 500 mL

Potassium Standard, 100 ppm APHA, Reagent, use for Water analysis. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Potassium Standard, 100 ppm APHA
Packaging Poly Bottle
CAS 7732-18-5, 7447-40-7
Quantity 500 mL
Color Colorless
For Use With (Equipment) Water Analyzer
Grade Reagent
Boiling Point 100°C
Specific Gravity 1.001
For Use With (Application) Water Analysis
Solubility Completely miscible
Physical Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.