Learn More
Sino Biological PPBP Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90132C08Y100
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
A DNA sequence encoding the cynomolgus / rhesus CXCL7 (XP_005555138.1) (Ser35-Asp128) was expressed with a polyhistidine tag at the C-terminus. Cynomolgus and Rhesus CXCL7 sequences are identical.
Catalog No. 50-237-2953