Learn More
Biotang Inc Accuraci* Disposable Reagent Reservoirs,100ml, 5/pk, 200/cs, sterile. Disposable reservoirs hold solutions for multichannel pipetting. Made of virgin modified polystyrene and accommodate all types of pipettors up to 12-channel widt