Learn More
Biotang Inc Accuraci* Disposable Reagent Reservoirs,50ml, 5/pk, 200/cs, sterile. Disposable reservoirs hold solutions for multichannel pipetting. Made of virgin modified polystyrene and accommodate all types of pipettors up to 12-channel widt
Supplier:  Biotang Inc 90105
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Accuraci* Disposable Reagent Reservoirs,50ml, 5/pk, 200/cs, sterile. Disposable reservoirs hold solutions for multichannel pipetting. Made of virgin modified polystyrene and accommodate all types of pipettors up to 12-channel widt
Catalog No. 50-751-5031