Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pramipexole dihydrochloride, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Dopamine receptor agonist exhibiting selectivity for the D3 receptor (Ki values are 0.5, 3.3, 3.9 and 3.9 nM for D3, D2S, D2L and D4 receptors respectively). Exhibits negligible affinity for D1 and D5 receptors. Displays activity in the treatment of Parkinson's disease.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 284.25 |
Percent Purity | >99% |
Chemical Name or Material | Pramipexole dihydrochloride |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction