Learn More
TARGETMOL CHEMICALS INC PRANIDIPINE 10MG
Supplier:  TARGETMOL CHEMICALS INC T16571
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pranidipine (OPC-13340) is a novel long-acting 14-dihydropyridine calcium channel blocker. It prolongs acetylcholine-induced relaxation in presence of endothelium as well as nitroglycerin-induced relaxation in absence of endothelium. It also has antihypertensive activity. purity: 99%
Catalog No. 50-257-7331
missing translation for 'QuantityLeft'