Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Praseodymium(III) fluoride, anhydrous, 99.9% (metals basis), Thermo Scientific™

Catalog No. p-7051519
Click to view available options
Quantity:
10 g
50 g

For vacuum deposition

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 13709-46-1
Molecular Formula F3Pr
Molecular Weight (g/mol) 197.90
MDL Number MFCD00016287
InChI Key BOTHRHRVFIZTGG-UHFFFAOYSA-K
IUPAC Name praseodymium(3+) trifluoride
SMILES [F-].[F-].[F-].[Pr+3]

Specifications

Melting Point 1395°C
Color Green
Physical Form Pieces
Quantity 10 g
UN Number UN3288
Solubility Information Soluble moderately in strong mineral acids Insoluble in water.
Formula Weight 197.9
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Density 6.2 g/mL
Assay (metals basis)
Chemical Name or Material Praseodymium(III) fluoride, Anhydrous
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 237-254-9
TSCA Yes
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.