Learn More
MilliporeSigma™ Supelco™ Premium Grade 304 Stainless Steel Tubing
Specially cleaned to ensure inertness; use for sensitive GC-MS applications
Supplier:  MilliporeSigma™ Supelco™ 20552
 View more versions of this product
Catalog No. 11-100-3738
Description
Description
  • Type 304 Stainless Steel is covered by highly protective film of Chromium Oxyhydroxide
  • It is recommended to add inhibitors like heterocyclic compounds to reduce the corrosion rate
  • 304SS materials are suitable for tubing and for wires as they exhibit strength and are corrosion resistant
  • Specially cleaned to ensure inertness
  • Use for sensitive GC-MS applications
Product Suggestions
Product Suggestions