Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Premium Grade Leather Drivers Gloves

Catalog No. p-9458005 Shop All Protective Industrial Products Inc. Products
Click to view available options
Size:
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Gloves provide abrasion resistance and dexterity.

  • Ideal for construction, machine operation, utility work, farming, roping, and other general applications
  • Top-grain leather
  • Straight thumb
  • Launderable

Specifications

Color Natural
Grade Premium
Material Top Grain Cowhide Leather
Product Type Driver Gloves
Size Medium
For Use With (Application) Construction, Machine Operation, Light Duty Welding, Utility Work, Farming, Roping, and Many Other General Applications
Glove Cuff Style Open
Hem Color Green
Thumb Style Straight
Length (English) 9.3 in.
Length (Metric) 23.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.