Learn More
TARGETMOL CHEMICALS INC PRIDOPIDINE 10MG
Supplier:  TARGETMOL CHEMICALS INC T7352
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pridopidine (FR310826) a dopamine (DA) stabilizer acts as a low affinity dopamine D2 receptor (D2R) antagonist improves motor performance and shows neuroprotective effects in Huntington disease R6/2 mouse model. purity: 99%
Catalog No. 50-257-6322
missing translation for 'QuantityLeft'