Learn More
Gold Biotechnology Inc ProBlock™ Gold Bacterial Protease Inhibitor Cocktail [100X] 1 mL
SDP
Supplier:  Gold Biotechnology Inc GB3301
Encompass_Preferred
ProBlock™ Gold Bacterial Protease Inhibitor Cocktail contains optimized concentrations of various bacterial protease inhibitors, which provide excellent inhibition of protease activities during protein purification from bacteria. The cocktail contains both reversible and irreversible protease inhibitors to inhibit serine and cysteine proteases. It also contains specific inhibitors for bacterial proteases, such as aspartic proteases and aminopeptidases. A 1X concentration of ProBlock™ Gold Bacterial inhibits over 90% of protease activities.
Catalog No. NC0991044
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.