Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ergodyne™ ProFlex™ 924LTR Leather-Reinforced Hybrid Dorsal Impact-Reducing Gloves
Shop All Ergodyne Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Description
- Leather-reinforced fingers and thumb saddle
- Hi-vis breathable mesh construction
- Neoprene cuff with molded hook and loop closure keeps out debris
- Gloves feature ID space to write user's names
Specifications
Specifications
Cuff Material | Neoprene |
Color | Hi-vis Lime Green, Black |
Hand | Specific |
Material | Thermoplastic Rubber |
Product Type | Impact-Reducing Gloves |
Size | Small |
For Use With (Application) | Roofing, Mining, Oil/Gas Drilling, Construction, and Mechanics |
Glove Cuff Style | Hook and Loop |
Series | ProFlex™ 924LTR |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction