Learn More
STERIS™ ProKlenz Booster Detergent
ProKlenz Booster High Performance Detergent Additive is used for boosting the performance of both alkaline and acidic cleaners and for manual cleaning applications. Its surfactancy and oxidizing chemistry allow for a multitude of cleaning mechanisms. pH - undiluted 5.5, typical
Supplier:  STERIS™ 1609D9
 View more versions of this product
Catalog No. 17-183-979
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Description
Description
ProKlenz Booster High Performance Detergent Additive can resolve your most challenging process cleaning challenges by greatly enhancing the performance of other detergents with a state-of-the-art surfactant system and oxidizing chemistry.
HOW PROKLENZ BOOSTER WORKS
ProKlenz Booster Detergent Additive is a proprietary blend of surfactants, an oxidizing agent and other performance-enhancing ingredients that provide multiple cleaning mechanisms. This product removes a wide range of process residues and is ideal for use in CIP, COP, and manual applications.
WHY PROKLENZ BOOSTER?
  • Productivity - Enhances cleaning performance of other detergents or works individually with its superior blend of surfactants and oxidizing chemistry. An excellent product for tablet coating removal in pharmaceutical and dietary supplement manufacturing facilities.
  • Easy to Validate - Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports.
  • Cost Savings - Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics (above 50°C) when used in clean-in-place (CIP), clean-out-of-place (COP), and manual applications.
  • Globally Availability - phosphate free, and manufactured with biodegradable ingredients to support harmonization of cleaning processes.
  • Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility.
Physical Properties: pH - undiluted 5.5, typical
Specifications
Specifications
Detergent
Clear Liquid
Unscented
Tote
No
5.5 (Undiluted)
Non-sterile
300 gal.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.