Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega CAMK2gamma Kinase Enzyme System

Catalog No. PRV9201 Shop All Promega Corporation Products
Click to view available options
Quantity:
1/Ea.
10 μg
Product Type:
ADP-Glo Kinase Assay and CAMK2 gamma Kinase Enzyme System
CAMK2 gamma Kinase Enzyme System

Recombinant human CAMK2 gamma (C-terminal truncation) was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. CAMK2 gamma is a member of the CAMKII family.

  • Easily Screen and Profile CAMK2γ Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Product Type ADP-Glo Kinase Assay and CAMK2 gamma Kinase Enzyme System
Includes 1 x 10μg CAMK2gamma Kinase Enzyme system, 1 x 1000 ADP-Glo™ Kinase assay
Molecular Weight (g/mol) 60
Content And Storage Components at different temps
For Use With (Application) Implicated in diverse effects of hormones and neurotransmitters and both primary screening as well as Kinase selectivity profiling
Quantity 1/Ea.
Form Fluid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.