Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proprietary Solvent III-1, Anhydrous, BAKER ANALYZED™ Reagent, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
4L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle
Specifications
Specifications
Chemical Name or Material | Proprietary Solvent III-1 |
Name Note | Anhydrous |
Grade | BAKER ANALYZED™ Reagent |
Packaging | Narrow Mouth Amber Glass Bottle |
Specific Gravity | 1L = 0.79kg |
Flash Point | 13.00°C |
Quantity | 4L |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction