Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propylene carbonate, MP Biomedicals™

Catalog No. p-4405216 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
500 g
1 kg
5 kg

Chemical Identifiers

CAS 108-32-7
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.09
MDL Number MFCD00005385,MFCD00798264,MFCD00798265
InChI Key RUOJZAUFBMNUDX-UHFFFAOYNA-N
Synonym propylene carbonate, 1,2-propylene carbonate, 1,2-propanediol cyclic carbonate, texacar pc, 1,3-dioxolan-2-one, 4-methyl, cyclic propylene carbonate, arconate 5000, 1,2-propanediol carbonate, 1-methylethylene carbonate, cyclic 1,2-propylene carbonate
PubChem CID 7924
IUPAC Name 4-methyl-1,3-dioxolan-2-one
SMILES CC1COC(=O)O1

Specifications

Color Colorless
Density ∽1.19g/mL
Quantity 500 g
Formula Weight 102.09
Physical Form Liquid
Chemical Name or Material Propylene carbonate, For GC analysis
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.