Learn More
Enzo Life Sciences Proteasome 20S β5i subunit monoclonal antibody (LMP7-1) (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW88450100
Encompass_Preferred
Synonyms: Proteasome subunit β type-8, LMP7, RING10 protein, Macropain subunit C13. Clone: LMP7-1. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant human proteasome subunit β5i. UniProt ID: P28062. Source: Purified from ascites. Species reactivity: Human, Rat. Applications: IHC, WB. Purification: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.4, containing 10mM sodium azide.
Catalog No. 50-210-0539
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.