Learn More
3M™ Protecta™ Comfort Vest-Style Harness
Protecta™ Comfort Vest-Style Harness minimizes workday readjustment and is designed for comfort and convenience
Supplier: 3M™ 7012816693
Description
- Fixed D-ring reduces the need for readjustment throughout the workday
- Specifically designed for added comfort
- Moisture-wicking, breathable back padding with soft edging for added comfort
- Modern design gives harness a fresh and sleek look
- ANSI-compliant auto-resetting lanyard keepers for fast and easy parking of snap hooks and carabiners
- Impact indicators make it easy to inspect the harness for prior damaging impact loads
- Protected labels for long-term identification and inspection
Specifications
Pass-Thru; Parachute; Tongue Buckle | |
420 lb. | |
Polyester | |
Comfort Vest-Style Harness | |
Back |
190 kg | |
3600 lb./ANSI | |
X-Large | |
Cleaning, Painting, Electrical, Machine Operations, Sanding |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.