Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Protecta™ Vest-Style Retrieval Harness
3M™ Protecta™ Vest-Style Retrieval Harness features a fixed D-ring to minimize workday readjustment.
Supplier: 3M™ 7100184604
Description
- Replaces Manufacturer Part Number 1191241
- Fixed D-ring reduces the need for readjustment throughout the workday
- Modern design gives harness a fresh and sleek look
- ANSI-compliant auto-resetting lanyard keepers for fast and easy parking of snap hooks and carabiners
- Impact indicators make it easy to inspect the harness for prior damaging impact loads
- Protected labels for long-term identification and inspection
- Back and Shoulder D-rings, Tongue-Buckle Leg Connection, Pass-Through Chest Connection
Specifications
ANSI Z359.11, OSHA 1926.502 | |
3600 lb./ANSI | |
190 kg | |
Back, Shoulder | |
Medium/Large | |
3.44 lb. |
Polyester | |
Pass-through, Parachute, Tongue Buckle | |
420 lb. | |
Vest-Style Retrieval Harness | |
Electrical, Painting, Assembly, Machine Operations, Cleaning, Grinding, Blasting, Remediation, Demolition, Sanding, Facility Maintenance |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?