Learn More
Glas Col LLC Protective Shield
SDP
Supplier:  Glas Col LLC 108C SHG1
Encompass_Preferred
Splash-Guard is a simple lab helper and personal protective equipment (PPE). It is a convenient way to provide anyone with a safer, well defined, comfortable work area. It provides a barrier against droplets and helps prevent the spreading of micro-organisms to your work area. Position several of the shields at intervals along your work bench to keep different set ups from encroaching on each other. Guard your project from the mishaps of nearby activities or personnel. 24" x 24"
Catalog No. NC1830924
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.