Learn More
Gold Biotechnology Inc Protein A Agarose Resin (Pharmaceutical grade for research) 100 mL
SDP
Supplier:  Gold Biotechnology Inc P400100
Encompass
Protein A agarose resin consists of rProtein A covalently bound to cross-linked agarose beads. It provides a very stable bond that can greatly minimize leakage of the Protein A allowing for reuse of the affinity resin in several purification steps. Protein A Agarose resin is available in bulk from 5-100 ml quantities.
Catalog No. NC2067695
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.