Learn More
Bioworld Protein-A Gold™ (5-10 nm), 1 mL
Supplier:  Bioworld 201000011
Encompass_Preferred
Protein-A Gold Protein-A is a highly stable cell surface receptor produced by several strains of Staphylococcus aureus. It is capable of binding to the Fc portion of immunoglobulins, especially IgG from a large number of species. Protein-A may be conjugated with various reporter molecules, including colloidial gold, without affecting the antibody binding site on the molecule. Specifications:Particle size5-10 nm (monodisperse), nominal 15 nm (colloidal gold) Suspension in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin, 20% glycerol, and 15 mM sodium azide. Storage: 2-8 C
Catalog No. 50-197-472
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.