Learn More
Gold Biotechnology Inc Proteinase K, RNase/DNase free, 39450-01-6, MFCD00132129, 1 g
Supplier: Gold Biotechnology Inc P4801
MW: 28.5 kDa, Activity: ≥30 U/mg, Store at -20°C. Proteinase K is a highly reactive nonspecific serine protease that belongs to the subtilisin family of proteins.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.