Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Proteome Profiler Mouse Apoptosis Array
Shop All R&D Systems Products
Click to view available options
:
1 Kit
Description
Contains 8 membranes, each spotted in duplicate with 21 different apoptosis antibodies.
Specifications
Specifications
Content And Storage | Store the unopened product at 2 to 8 °C. Do not use past expiration date |
Quantity | 1 Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction