Learn More
Tocris Bioscience™ Proteome Profiler Rat XL Cytokine Array
GSA_VA
Catalog No. ARY030 Shop All R&D Systems Products
Click to view available options
:
Proteome Profiler Rat XL Cytokine Array
This item is not returnable. View return policy
This item is not returnable. View return policy
For parallel determination of relative levels of selected rat cytokines.
The Proteome Profiler Rat XL Cytokine Array contains 4 membranes-each spotted in duplicate with 79 different cytokine antibodies.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Description Four membranes-each spotted in duplicate with 79 different cytokine antibodies
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Kit Contents Array Buffer, Chemi Reagent, Detection Antibody Cocktail, Rat XL Cytokine Array, Rat XL Cytokine Array nitrocellulose membranes, Rectangular four-well multi-dish, Streptavidin-HRP, Transparency Overlay Template, Wash Buffer Concentrate (25X)
Product Type Capture Antigen ELISA kit
For Laboratory Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.