Learn More
Electron Microscopy Sciences PS4R with Graticule certificate
Supplier:  Electron Microscopy Sciences 680474RGR
Encompass_Preferred
These stage micrometers are available with a calibration certificate that provides traceability for the precise calibration and confirmation of accuracy of optical measuring instruments, necessary under ISO provisions. The glass discs are mounted in stainless steel slides with engraved serial numbers. Each slide is supplied in a polished wooden storage case. A Certificate of Comparison from Pyser-SGI compares the scale to a NPL calibrated in-house standards. Reflected light stage micrometer with 0.1" scale subdivided to 0.001".
Catalog No. 50-190-1564
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.