Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSB 0739, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Highly potent P2Y12 receptor antagonist (Ki = 24.9 nM). Unlike clopidogrel (Cat. No. 2490), does not require bioactivation.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 609.54 |
Percent Purity | >96% |
Chemical Name or Material | PSB 0739 |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction