Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSB 0777 ammonium salt, Tocris Bioscience™

Catalog No. p-4816533 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent adenosine A2A agonist

Potent adenosine A2A receptor full agonist (Ki = 44.4 nM for rat brain striatal A2A receptors). Exhibits subtype selectivity for A2A receptors over A1, A2B and A3 receptors (Ki ≥ 10 000 nM for rat A1, human A2B and human A3 receptors). Potentiates acetylcholine-induced contractions in ex vivo rat ileum/jejunum segments.

Chemical Identifiers

CAS 2122196-16-9
Molecular Formula C18H24N6O7S2
Molecular Weight (g/mol) 500.55
InChI Key AYQGFWKUGKSGCZ-UHFFFAOYNA-N
Synonym psb 0777 ammonium salt, 4-2-6-amino-9-b-d-ribofuranosyl-9h-purin-2-yl thio ethyl benzenesulfonic acid ammonium salt
PubChem CID 90488932

Specifications

Quantity 10 mg
Formula Weight 500.55
Percent Purity >98%
Chemical Name or Material PSB 0777 ammonium salt
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.