Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSB 0788, Tocris Bioscience™
GSA_VA

Catalog No. p-4784361 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective A2B antagonist

Potent and selective adenosine A2B receptor antagonist (IC50 = 3.64 nM at human A2B receptors). Displays > 100-fold selectivity over hA1, hA2A and hA3 receptors.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 1027513-54-7
Molecular Formula C25H27ClN6O4S
Molecular Weight (g/mol) 543.039
InChI Key JQZJACVYMPKVDS-UHFFFAOYSA-N
Synonym d05xgf, 8-4-4-4-chlorobenzyl piperazine-1-sulfonyl phenyl-1-propylxanthine, 8-4-4-4-chlorobenzyl piperazide-1-sulfonyl phenyl-1-propylxanthine, 8-4-4-4-chlorobenzyl piperazin-1-ylsulfonyl phenyl-1-propyl-1h-purine-2,6 3h,7h-dione, 8-4-4-4-chlorophenyl methyl piperazine-1-sulfonyl phenyl-1-propyl-2,3,6,7-tetrahydro-1h-purine-2,6-dione
PubChem CID 44186581
IUPAC Name 8-[4-[4-[(4-chlorophenyl)methyl]piperazin-1-yl]sulfonylphenyl]-1-propyl-3,7-dihydropurine-2,6-dione
SMILES CCCN1C(=O)C2=C(NC1=O)N=C(N2)C3=CC=C(C=C3)S(=O)(=O)N4CCN(CC4)CC5=CC=C(C=C5)Cl

Specifications

Quantity 10 mg
Formula Weight 543.04
Percent Purity >98%
Chemical Name or Material PSB 0788
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.