Learn More
Pseudomonas aeruginosa Mouse anti-Bacteria, DyLight 350, Clone: 1001/214, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP125946UV
Catalog No. NBP125946UV
Add to Cart
Description
Description
Pseudomonas aeruginosa Monoclonal antibody specifically detects Antigen in Bacteria samples. It is validated for Enzyme Linked Immunosorbent Assay, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Pseudomonas Aeruginosa
1001/214
ELISA, Immunocytochemistry/Immunofluorescence
Purified
Mouse
IgG3
0.1 ml
Store at 4C in the dark.
Monoclonal
Bacteria
ELISA, Immunocytochemistry, Immunofluorescence
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
Pseudomonas aeruginosa 9
P. aeruginosa serotype 3 (ATCC 33350)
Protein A purified
RUO
Primary
NBP1-25946 is specific for P. aeruginosa serotype 3.
Product Suggestions
Product Suggestions