Learn More
Pseudomonas aeruginosa Mouse anti-Bacteria, DyLight 550, Clone: 1001/214, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP125946R
Description
Pseudomonas Aeruginosa Monoclonal specifically detects Pseudomonas Aeruginosa in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Pseudomonas Aeruginosa | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein A purified | |
RUO | |
NBP1-25946 is specific for P. aeruginosa serotype 3. | |
Store at 4C in the dark. | |
IgG3 |
ELISA, Immunocytochemistry, Immunofluorescence | |
1001/214 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
Pseudomonas aeruginosa 9 | |
P. aeruginosa serotype 3 (ATCC 33350) | |
0.1 mL | |
Primary | |
Bacteria | |
Purified |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.