Learn More
Gold Biotechnology Inc AGL-1 (pSOUP-P19) Agrobacterium Electrocompetent Cells 5x50 µl
SDP
Supplier:  Gold Biotechnology Inc CC2285X50
Encompass_Preferred
AGL-1 (pSoup-P19) electrocompetent Agrobacterium cells are a highly efficient system for genetic transformation in various plant species. This strain is derived from the C58 RecA-type Agrobacterium strain and carries the rifampicin, carbenicillin, and streptomycin resistance genes for selection purposes. The Ti plasmid pTiBo542DT-DNA used in this system has a deleted T-DNA region, and instead contains a binary vector that restores the T-DNA binary system function.
Catalog No. NC2942867
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.