Learn More
Medchemexpress LLC HY-N0828 25mg Medchemexpress, Pterostilbene CAS:537-42-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN082825MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Medchemexpress, HY-N0828 25mg Pterostilbene CAS:537-42-8 Pterostilbene is a stilbenoid isolated from blueberries and Pterocarpus marsupium [1] . Shows anti-oxidant, anti-inflammatory, anti-carcinogenic, anti-diabetic and anti-obesity properties [1] [4] . Pterostilbene blocks ROS production [3] , also exhibits inhibitory activity against various free radicals such as DPPH, ABTS, hydroxyl, superoxide and hydrogen peroxide [4] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3734
missing translation for 'QuantityLeft'