Learn More
BEZWADA BIOMEDICAL LLC Poly(trimethylene carbonate-co-D,L-lactide), TMC:D,L-lactide 50:50 viscosity 1.0 to 2.0 dl/g, 5.0 GM
SDP
Supplier:  BEZWADA BIOMEDICAL LLC 5455
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Polytrimethylene carbonate amorphous rubbery polymer. Copolymers of Trimethylene carbonate is prepared for varying absorption profile, stiffness and crsytallinity. The polymer is elastomeric. Polymers are used in drug delivery and medical device coatings.
Catalog No. 50-190-4499