Learn More
Volu Sol Pure Ethanol Absolute (200 Proof), Tax-Paid -(128 oz / 3.78 L)-Case of 4
Pure Ethyl Alcohol, 200 Proof. Meets ACS Specifications, For nucleic acid purification and precipitation, Ultrapure molecular biology grade ethanol is used for the purification and precipitation of biomolecules such as nucleic acids and proteins, It is also used in histology to prepare staining and destaining reagents and for dehydrating tissues prior to embedding. Price includes Federal Excise Tax