Learn More
MilliporeSigma™ Supelco™ Purge/Trap A
Specific configurations available for a variety of instruments
Supplier:  MilliporeSigma™ Supelco™ 21059U
 View more versions of this product
Catalog No. 11-101-6254
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Description
Description
  • Tubes used in purge and trap analyses generally are packed with multiple beds of adsorbent materials
  • A single tube can trap a broad range of polar and nonpolar, high and low molecular weight compounds
  • Each bed protects the next, increasingly active bed, by preventing compounds from being held so strongly that they cannot be desorbed quickly without decomposition
  • During the purge phase of sampling, lower molecular weight compounds pass through the initial adsorbent beds but are trapped by succeeding beds
  • During desorption, the carrier gas passes through the trap in the reverse direction of purge flow, so that higher molecular weight compounds never come in contact with the stronger (innermost) adsorbents
Specifications
Specifications
Purge/Trap A
Tekmar™ Velocity XPT, LSC-1, LSC-2, LSC-2000, 4000 (Straight, 12 in. Swagelok Fitting), HT3
Industrial Hygiene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.