Learn More
Biolegend Purified anti-PPM1B
Supplier:  Biolegend 934201
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
Purified anti-PPM1B [W19264A]; Isotype: Rat IgG2b, κ; Reactivity: Human, Mouse; Apps: WB, KO/KD-WB; Size: 25 μg
Catalog No. 50-272-1610
missing translation for 'QuantityLeft'