Learn More
Enzo Life Sciences SCREEN-WELL Purinergic ligand library (100µl/well)
SDP
Supplier:  Enzo Life Sciences BML28200100
Encompass_Preferred
The SCREEN-WELL Purinergic and Adenosine Ligands Plate contains 72 CNS receptor ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The Purinergic and Adenosine Ligands Plate is 1 plate of the larger Neurotransmitter Library (BML-2810) which contains over 650 ligands. Long Term Storage: -80°C. Applications: HTS .
Catalog No. 50-201-0206
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.